Fmoc-6-Ahx-OH 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Fmoc-ε-aminocaproic acid, Fmoc-ε-Acp-OH, 6-(Fmoc-amino)hexanoic acid
H63148-03
H63148-06
H63148-14
AAH63148-03EA
58.14
CAD
AAH63148-03
AAH63148-06
AAH63148-14
Fmoc-6-Ahx-OH 98%
Fmoc-6-Ahx-OH
6-(Fmoc-amino)hexanoic acid, 98%
Purity: 98%
CAS No.: 88574-06-5
Molecular Formula: C21H23NO4
Formula Weight: 353.41
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 5883220
MDL No.: MFCD00077045
Formula:
C₂₁H₂₃NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00077045 CAS Number: 88574-06-5 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...