Ethyl-1-boc-piperazine-3-carboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63146-03
H63146-06
AAH63146-03EA
369.72
CAD
AAH63146-03
AAH63146-06
AAH63146-MD
Ethyl-1-boc-piperazine-3-carboxylate 97%
Ethyl-1-boc-piperazine-3-carboxylate
Ethyl 1-Boc-piperazine-3-carboxylate, 97%
Purity: 97%
CAS No.: 183742-29-2
Molecular Formula: C12H22N2O4
Formula Weight: 258.31
Physical Form: Solid
MDL No.: MFCD13172266
Formula:
C₁₂H₂₂N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD13172266 CAS Number: 183742-29-2 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...