Methyl-2-bromo-4-methylbenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63144-03
H63144-06
AAH63144-03EA
133.35
CAD
AAH63144-03
AAH63144-06
AAH63144-MD
Methyl-2-bromo-4-methylbenzoate 98%
Methyl-2-bromo-4-methylbenzoate
Methyl 2-bromo-4-methylbenzoate, 98%
Purity: 98%
CAS No.: 87808-49-9
Molecular Formula: C9H9BrO2
Formula Weight: 229.07
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD03840737
Formula:
C₉H₉BrO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD03840737 CAS Number: 87808-49-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...