2-Bromo-L-phenylalanine 95%
Supplier: Thermo Scientific Chemicals
H63141-03
H63141-06
AAH63141-03EA
134.93
CAD
AAH63141-03
AAH63141-06
AAH63141-MD
2-Bromo-L-phenylalanine 95%
2-Bromo-L-phenylalanine
2-Bromo-L-phenylalanine, 95%
Purity: 95%
CAS No.: 42538-40-9
Molecular Formula: C9H10BrNO2
Formula Weight: 244.09
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Off-white
MDL No.: MFCD03092958
Formula:
C₉H₁₀BrNO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD03092958 CAS Number: 42538-40-9 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...