3-(3-Pyridyl)-D-alanine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63132-03
H63132-06
AAH63132-03EA
237
CAD
AAH63132-03
AAH63132-06
AAH63132-MD
3-(3-Pyridyl)-D-alanine 95%
3-(3-Pyridyl)-D-alanine
3-(3-Pyridyl)-D-alanine, 95%
Purity: 95%
CAS No.: 70702-47-5
Molecular Formula: C8H10N2O2
Formula Weight: 166.18
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Off-white
Beilstein Reference: 4351584
MDL No.: MFCD00079673
Formula:
C₈H₁₀N₂O₂ MW: 202.638 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00079673 CAS Number: 70702-47-5 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...