3-Fluoro-D-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

H63131-03 H63131-06
AAH63131-03EA 104.28 CAD
AAH63131-03 AAH63131-06 AAH63131-14
3-Fluoro-D-phenylalanine 95%
3-Fluoro-D-phenylalanine

3-Fluoro-D-phenylalanine, 95%


Purity: 95% CAS No.: 110117-84-5 Molecular Formula: C9H10FNO2 Formula Weight: 183.18 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD00066449

Formula: C₉H₁₀FNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00066449
CAS Number: 110117-84-5

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR