5-Methoxy-7-methyl-1H-indole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63123-03
H63123-06
AAH63123-03EA
295.46
CAD
AAH63123-03
AAH63123-06
AAH63123-MD
5-Methoxy-7-methyl-1H-indole 97%
5-Methoxy-7-methyl-1H-indole
5-Methoxy-7-methylindole, 97%
Purity: 97%
CAS No.: 61019-05-4
Molecular Formula: C10H11NO
Formula Weight: 161.2
Physical Form: Solid
MDL No.: MFCD09027022
Formula:
C₁₀H₁₁NO Storage Temperature: Ambient |
MDL Number:
MFCD09027022 CAS Number: 61019-05-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...