DL-Homoserine 98%
Supplier: Thermo Scientific Chemicals
H63118-03
H63118-06
AAH63118-03EA
99.22
CAD
AAH63118-03
AAH63118-06
AAH63118-MD
DL-Homoserine 98%
DL-Homoserine
DL-Homoserine, 98%
Purity: 98%
CAS No.: 1927-25-9
Molecular Formula: C4H9NO3
Formula Weight: 119.12
Physical Form: Solid
Melting Point: 188-189°
Beilstein Reference: 1721682
MDL No.: MFCD00002618
Formula:
C₄H₉NO₃ Melting Pt: 188…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002618 CAS Number: 1927-25-9 EINECS: 217-661-8 Merck Index: 14,4739 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...