Methyl-3-cyanoindole-6-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-cyano-1H-indole-6-carboxylate

H63100-03 H63100-06 H63100-MD
AAH63100-03EA 317.58 CAD
AAH63100-03 AAH63100-06 AAH63100-MD
Methyl-3-cyanoindole-6-carboxylate 97%
Methyl-3-cyanoindole-6-carboxylate

Methyl 3-cyanoindole-6-carboxylate, 97%


Purity: 97% CAS No.: 1000576-51-1 Molecular Formula: C11H8N2O2 Formula Weight: 200.19 Physical Form: Solid MDL No.: MFCD09878564

Formula: C₁₁H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09878564
CAS Number: 1000576-51-1

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR