4-Aminocarbethoxypiperidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Ethoxycarbonylamino)piperidine

H63095-03 H63095-06
AAH63095-03EA 137.93 CAD
AAH63095-03 AAH63095-06 AAH63095-MD
4-Aminocarbethoxypiperidine 97%
4-Aminocarbethoxypiperidine

4-(Ethoxycarbonylamino)piperidine, 97%


Purity: 97% CAS No.: 64951-36-6 Molecular Formula: C8H16N2O2 Formula Weight: 172.22 Physical Form: Solid MDL No.: MFCD03844679

Formula: C₈H₁₆N₂O₂
MW: 172.228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03844679
CAS Number: 64951-36-6

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR