Ethyl-4,6-dichloronicotinate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63092-03
H63092-06
AAH63092-03EA
109.49
CAD
AAH63092-03
AAH63092-06
AAH63092-MD
Ethyl-4,6-dichloronicotinate 95%
Ethyl-4,6-dichloronicotinate
Ethyl 4,6-dichloronicotinate, 95%
Purity: 95%
CAS No.: 40296-46-6
Molecular Formula: C8H7Cl2NO2
Formula Weight: 220.05
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Appearance: Yellow
MDL No.: MFCD00234408
Formula:
C8H7Cl2NO2 Storage Temperature: Refrigerator |
MDL Number:
MFCD00234408 CAS Number: 40296-46-6 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...