1-Boc-4-(aminomethyl)-4-hydroxypiperidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63079-03
H63079-06
AAH63079-03EA
129.09
CAD
AAH63079-03
AAH63079-06
AAH63079-MD
1-Boc-4-(aminomethyl)-4-hydroxypiperidine 97%
1-Boc-4-(aminomethyl)-4-hydroxypiperidine
4-Aminomethyl-1-Boc-4-hydroxypiperidine, 97%
Purity: 97%
CAS No.: 392331-66-7
Molecular Formula: C11H22N2O3
Formula Weight: 230.3
Physical Form: Solid
Sensitivity: Air Sensitive
Appearance: Off-white
MDL No.: MFCD11579803
Formula:
C₁₁H₂₂N₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD11579803 CAS Number: 392331-66-7 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...