N-(tert-Butoxycarbonyl)-L-phenylalanine methyl ester 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63078-06
H63078-14
AAH63078-06EA
54.19
CAD
AAH63078-06
AAH63078-14
AAH63078-22
N-(tert-Butoxycarbonyl)-L-phenylalanine methyl ester 95%
N-(tert-Butoxycarbonyl)-L-phenylalanine methyl ester
N-Boc-L-phenylalanine methyl ester, 95%
Purity: 95%
CAS No.: 51987-73-6
Molecular Formula: C15H21NO4
Formula Weight: 379.33
Flash Point: 113°(235°F)
Physical Form: Powder
Appearance: Off-white
Melting Point: 36-40°
Beilstein Reference: 3061910
Optical Rotation: -4°(c=2 in methanol)
MDL No.: MFCD00076977
Formula:
C₁₅H₂₁NO₄ Melting Pt: 36…40 °C Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00076977 CAS Number: 51987-73-6 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...