DL-Glutamic acid 95%

Supplier: Thermo Scientific Chemicals

H63075-14 H63075-22 H63075-36
AAH63075-14EA 90.06 CAD
AAH63075-14 AAH63075-22 AAH63075-36
DL-Glutamic acid 95%
DL-Glutamic acid

DL-Glutamic acid, 95%


Purity: 95% CAS No.: 617-65-2 Molecular Formula: C5H9NO4 Formula Weight: 147.13 Storage Temperature: -30°C to -10°C Physical Form: Solid Beilstein Reference: 1723799 MDL No.: MFCD00063113

Formula: C₅H₉NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00063113
CAS Number: 617-65-2
EINECS: 210-522-2
Merck Index: 14,4469

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR