Fmoc-O-methyl-D-tyrosine 98%
Supplier: Thermo Scientific Chemicals
H63073-03
H63073-06
AAH63073-03EA
82.32
CAD
AAH63073-03
AAH63073-06
AAH63073-MD
Fmoc-O-methyl-D-tyrosine 98%
Fmoc-O-methyl-D-tyrosine
N-Fmoc-O-methyl-D-tyrosine, 98%
Purity: 98%
CAS No.: 201335-88-8
Molecular Formula: C25H23NO5
Formula Weight: 417.45
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00237393
Formula:
C₂₅H₂₃NO₅ Storage Temperature: Ambient |
MDL Number:
MFCD00237393 CAS Number: 201335-88-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...