2-Iodopyridine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63062-06
H63062-14
AAH63062-06EA
120.24
CAD
AAH63062-06
AAH63062-14
2-Iodopyridine 95%
2-Iodopyridine
2-Iodopyridine, 95%
Purity: 95%
CAS No.: 5029-67-4
Molecular Formula: C5H4IN
Formula Weight: 205
Physical Form: Solid
Density: 1.928
Refractive Index: 1.632
Sensitivity: Light Sensitive
Appearance: Pale yellow
Melting Point: 52° dec
Beilstein Reference: 105790
MDL No.: MFCD00464928
Formula:
C₅H₄IN Melting Pt: 52 °C (dec.) Density: 1.928 Storage Temperature: Ambient |
MDL Number:
MFCD00464928 CAS Number: 5029-67-4 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...