N-Fmoc-N-methyl-D-phenylalanine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-D-N-Me-Phe-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-phenylpropanoic acid
H63055-03
H63055-06
AAH63055-03EA
232.26
CAD
AAH63055-03
AAH63055-06
AAH63055-MD
N-Fmoc-N-methyl-D-phenylalanine 98%
N-Fmoc-N-methyl-D-phenylalanine
N-Fmoc-N-methyl-D-phenylalanine, 98%
Purity: 98%
CAS No.: 138775-05-0
Molecular Formula: C25H23NO4
Formula Weight: 401.45
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 8441627
MDL No.: MFCD00153392
Formula:
C₂₅H₂₃NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00153392 CAS Number: 138775-05-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...