Cbz-L-ornithine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Z-Orn-OH, Nα-Benzyloxycarbonyl-L-ornithine, (S)-5-Amino-2-(((benzyloxy)carbonyl)amino)pentanoic acid, N-α-Cbz-L-Ornithine
H63052-03
H63052-06
AAH63052-03EA
57.2
CAD
AAH63052-03
AAH63052-06
Cbz-L-ornithine 98%
Cbz-L-ornithine
Nalpha-Benzyloxycarbonyl-L-ornithine, 98%
Purity: 98%
CAS No.: 2640-58-6
Molecular Formula: C13H18N2O4
Formula Weight: 266.29
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00038200
Formula:
C₁₃H₁₈N₂O₄ MW: 266.297 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00038200 CAS Number: 2640-58-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...