Fmoc-Lys(Dnp)-OH 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Nepsilon-2,4-Dinitrophenyl-Nα-Fmoc-L-lysine

H63050-03 H63050-06
AAH63050-03EA 227.52 CAD
AAH63050-03 AAH63050-06 AAH63050-MD
Fmoc-Lys(Dnp)-OH 98%
Fmoc-Lys(Dnp)-OH

N^e-2,4-Dinitrophenyl-N^a-Fmoc-L-lysine, 98%


Purity: 98% CAS No.: 148083-64-1 Molecular Formula: C27H26N4O8 Formula Weight: 534.52 Physical Form: Solid MDL No.: MFCD00273454

Formula: C₂₇H₂₆N₄O₈
MW: 534.525 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00273454
CAS Number: 148083-64-1

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR