Methyl-3-amino-5-fluorobenzoate 98%

Supplier: Thermo Scientific Chemicals

H63038-03 H63038-06 H63038-MD
AAH63038-03EA 135.88 CAD
AAH63038-03 AAH63038-06 AAH63038-MD
Methyl-3-amino-5-fluorobenzoate 98%
Methyl-3-amino-5-fluorobenzoate

Methyl 3-amino-5-fluorobenzoate, 98%


Purity: 98% CAS No.: 884497-46-5 Molecular Formula: C8H8FNO2 Formula Weight: 169.16 Physical Form: Solid Appearance: White to yellow MDL No.: MFCD05863683

Formula: C8H8FNO2
Storage Temperature: Ambient
MDL Number: MFCD05863683
CAS Number: 884497-46-5

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR