(S)-2-((tert-Butoxycarbonyl)amino)-3-(pyridin-2-yl)propanoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63037-03
H63037-06
AAH63037-03EA
132.72
CAD
AAH63037-03
AAH63037-06
AAH63037-MD
(S)-2-((tert-Butoxycarbonyl)amino)-3-(pyridin-2-yl)propanoic acid 98%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(pyridin-2-yl)propanoic acid
N-Boc-3-(2-pyridyl)-L-alanine, 98%
Purity: 98%
CAS No.: 71239-85-5
Molecular Formula: C13H18N2O4
Formula Weight: 266.29
Physical Form: Solid
Beilstein Reference: 6416850
Optical Rotation: -16°(c=1 in methanol)
MDL No.: MFCD00191190
Formula:
C13H18N2O4 Storage Temperature: Ambient |
MDL Number:
MFCD00191190 CAS Number: 71239-85-5 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...