3-Nitro-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-2-Amino-3-(3-nitrophenyl)propanoic acid, H-Phe(3-NO₂)-OH

H63029-03 H63029-06 H63029-MD
AAH63029-03EA 211.72 CAD
AAH63029-03 AAH63029-06 AAH63029-MD
3-Nitro-L-phenylalanine 95%
3-Nitro-L-phenylalanine

3-Nitro-L-phenylalanine, 95%


Purity: 95% CAS No.: 19883-74-0 Molecular Formula: C9H10N2O4 Formula Weight: 210.19 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: Off-white MDL No.: MFCD01860661

Formula: C₉H₁₀N₂O₄
MW: 210.189 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01860661
CAS Number: 19883-74-0

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR