S-Trityl-D-cysteine 98%
Supplier: Thermo Scientific Chemicals
H63022-03
H63022-06
AAH63022-03EA
87.69
CAD
AAH63022-03
AAH63022-06
S-Trityl-D-cysteine 98%
S-Trityl-D-cysteine
(-)-S-Trityl-D-cysteine, 98%
Purity: 98%
CAS No.: 25840-82-8
Molecular Formula: C22H21NO2S
Formula Weight: 363.47
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD00236948
Formula:
C₂₂H₂₁NO₂S Storage Temperature: Ambient |
MDL Number:
MFCD00236948 CAS Number: 25840-82-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...