2-Fluoro-6-nitrobenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Fluoro-2-nitrobenzoic acid
H63013-06
H63013-14
H63013-22
AAH63013-06EA
86.11
CAD
AAH63013-06
AAH63013-14
AAH63013-22
2-Fluoro-6-nitrobenzoic acid 98%
2-Fluoro-6-nitrobenzoic acid
2-Fluoro-6-nitrobenzoic acid, 98%
Purity: 98%
CAS No.: 385-02-4
Molecular Formula: C7H4FNO4
Formula Weight: 185.11
Physical Form: Solid
MDL No.: MFCD01862079
Formula:
C₇H₄FNO₄ MW: 185.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01862079 CAS Number: 385-02-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...