3-phenylimidazolidine-2,4-dione 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63009-03
H63009-MD
AAH63009-03EA
248.06
CAD
AAH63009-03
AAH63009-MD
3-phenylimidazolidine-2,4-dione 95%
3-phenylimidazolidine-2,4-dione
3-Phenylhydantoin, 95%
Purity: 95%
CAS No.: 2221-13-8
Molecular Formula: C9H8N2O2
Formula Weight: 176.17
Physical Form: Solid
Appearance: White
MDL No.: MFCD00209486
Formula:
C₉H₈N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00209486 CAS Number: 2221-13-8 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...