DL-tert-Leucine (H-DL-Tle-OH) 95%

Supplier: Thermo Scientific Chemicals

H63007-03 H63007-06
AAH63007-03EA 82.95 CAD
AAH63007-03 AAH63007-06
DL-tert-Leucine (H-DL-Tle-OH) 95%
DL-tert-Leucine (H-DL-Tle-OH)

DL-tert-Leucine, 95%


Purity: 95% CAS No.: 33105-81-6 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Physical Form: Solid Melting Point: 300° dec Beilstein Reference: 1721823 MDL No.: MFCD00065933

Formula: C₆H₁₃NO₂
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00065933
CAS Number: 33105-81-6

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR