Fmoc-O-(benzylphospho)-L-threonine 97%

Supplier: Thermo Scientific Chemicals

H63006-03 H63006-MD
AAH63006-03EA 459.78 CAD
AAH63006-03 AAH63006-MD
Fmoc-O-(benzylphospho)-L-threonine 97%
Fmoc-O-(benzylphospho)-L-threonine

N-Fmoc-O-benzylphospho-L-threonine, 97%


Purity: 97% CAS No.: 175291-56-2 Molecular Formula: C26H26NO8P Formula Weight: 511.46 Storage Temperature: 2°C to 8°C Physical Form: Solid Sensitivity: Moisture Sensitive Beilstein Reference: 7557104 MDL No.: MFCD00797870

Formula: C₂₆H₂₆NO₈P
Storage Temperature: Ambient
MDL Number: MFCD00797870
CAS Number: 175291-56-2

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR