6-Methoxy-1,2,3,4-tetrahydroquinoline 97%

Supplier: Thermo Scientific Chemicals
Warning

H63002-03 H63002-06 H63002-MD
AAH63002-03EA 145.04 CAD
AAH63002-03 AAH63002-06 AAH63002-MD
6-Methoxy-1,2,3,4-tetrahydroquinoline 97%
6-Methoxy-1,2,3,4-tetrahydroquinoline

6-Methoxy-1,2,3,4-tetrahydroquinoline, 97%


Purity: 97% CAS No.: 120-15-0 Molecular Formula: C10H13NO Formula Weight: 163.22 Physical Form: Solid Appearance: Yellow to brown Melting Point: 34-37° MDL No.: MFCD00090494

Formula: C₁₀H₁₃NO
Melting Pt: 34…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00090494
CAS Number: 120-15-0
EINECS: 204-374-8

Order Now

Specification Test Results

Assay 97%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR