H-Lys(boc)-OtBu·HCl 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride, H-Lys(Boc)-OtBu.HCl, H-Lys(Boc)-OtBu·HCl

H62993-03 H62993-06
AAH62993-03EA 159.58 CAD
AAH62993-03 AAH62993-06 AAH62993-MD
H-Lys(boc)-OtBu·HCl 95%
H-Lys(boc)-OtBu·HCl

N^e-Boc-L-lysine tert-butyl ester hydrochloride, 95%


Purity: 95% CAS No.: 13288-57-8 Molecular Formula: C15H30N2O4.HCl Formula Weight: 338.87 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White MDL No.: MFCD00038899

Formula: C₁₅H₃₁ClN₂O₄
MW: 338.873 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00038899
CAS Number: 13288-57-8

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR