Fmoc-D-Dab(Boc)-OH 98%

Supplier: Thermo Scientific Chemicals

H62954-03 H62954-06 H62954-MD
AAH62954-03EA 244.9 CAD
AAH62954-03 AAH62954-06 AAH62954-MD
Fmoc-D-Dab(Boc)-OH 98%
Fmoc-D-Dab(Boc)-OH

(R)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid, 98%


Purity: 98% CAS No.: 114360-56-4 Molecular Formula: C24H28N2O6 Formula Weight: 440.49 Physical Form: Powder Appearance: White MDL No.: MFCD00798647

Formula: C₂₄H₂₈N₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00798647
CAS Number: 114360-56-4

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR