Fmoc-D-Orn(Boc)-OH 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Nδ-Boc-Nα-Fmoc-D-ornithine

H62930-03 H62930-06
AAH62930-03EA 99.07 CAD
AAH62930-03 AAH62930-06
Fmoc-D-Orn(Boc)-OH 95%
Fmoc-D-Orn(Boc)-OH

N^d-Boc-N^a-Fmoc-D-ornithine, 95%


Purity: 95% CAS No.: 118476-89-4 Molecular Formula: C25H30N2O6 Formula Weight: 454.52 Physical Form: Powder Appearance: Yellow MDL No.: MFCD00077065

Formula: C₂₅H₃₀N₂O₆
MW: 454.523 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077065
CAS Number: 118476-89-4

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR