Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-Pyr-OBzl

H62912-03 H62912-06 H62912-MD
AAH62912-03EA 128.93 CAD
AAH62912-03 AAH62912-06 AAH62912-MD
Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate 98%
Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate

Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%


Purity: 98% CAS No.: 113400-36-5 Molecular Formula: C17H21NO5 Formula Weight: 319.35 Physical Form: Powder Appearance: White MDL No.: MFCD04115781

Formula: C₁₇H₂₁NO₅
Storage Temperature: Ambient
MDL Number: MFCD04115781
CAS Number: 113400-36-5

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR