Nɑ,N-ε-Bis(tert-butoxycarbonyl)-L-lysine dicyclohexylammonium salt 98%
Supplier: Thermo Scientific Chemicals
H62785-14
H62785-22
AAH62785-14EA
91.01
CAD
AAH62785-14
AAH62785-22
Nɑ,N-ε-Bis(tert-butoxycarbonyl)-L-lysine dicyclohexylammonium salt 98%
Nalpha,N-epsilon-Bis(tert-butoxycarbonyl)-L-lysine dicyclohexylammonium salt
N^a,N^e-Di-Boc-L-lysine dicyclohexylammonium salt, 98%
Purity: 98%
CAS No.: 15098-69-8
Molecular Formula: C16H30N2O6.C12H23N
Formula Weight: 527.74
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White
Melting Point: 139-143°
Beilstein Reference: 6262378
MDL No.: MFCD00038892
Formula:
C₂₈H₅₃N₃O₆ Melting Pt: 139…143 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00038892 CAS Number: 15098-69-8 EINECS: 239-150-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...