1-(Triphenylmethyl)-L-histidine 98%
Supplier: Thermo Scientific Chemicals
H62779-03
H62779-06
AAH62779-03EA
85.64
CAD
AAH62779-03
AAH62779-06
1-(Triphenylmethyl)-L-histidine 98%
1-(Triphenylmethyl)-L-histidine
1-Trityl-L-histidine, 98%
Purity: 98%
CAS No.: 35146-32-8
Molecular Formula: C25H23N3O2
Formula Weight: 397.47
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Melting Point: 210° dec
Beilstein Reference: 767244
MDL No.: MFCD00153444
Formula:
C₂₅H₂₃N₃O₂ Melting Pt: 210 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00153444 CAS Number: 35146-32-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...