Boc-Thr(tBu)-OH 95%
Supplier: Thermo Scientific Chemicals
H62713-03
H62713-06
AAH62713-03EA
64.46
CAD
AAH62713-03
AAH62713-06
Boc-Thr(tBu)-OH 95%
Boc-Thr(tBu)-OH
N-Boc-O-tert-butyl-L-threonine, 95%
Purity: 95%
CAS No.: 13734-40-2
Molecular Formula: C13H25NO5
Formula Weight: 275.34
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 95-98°
Beilstein Reference: 4454820
Optical Rotation: +3°(c=2 in DMF)
MDL No.: MFCD00151115
Formula:
C₁₃H₂₅NO₅ Melting Pt: 95…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151115 CAS Number: 13734-40-2 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...