(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid 97%
Supplier: Thermo Scientific Chemicals
H62705-06
H62705-14
AAH62705-06EA
89.59
CAD
AAH62705-06
AAH62705-14
AAH62705-22
(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid 97%
(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid
N-Boc-O-benzyl-L-serine, 97%
Purity: 97%
CAS No.: 23680-31-1
Molecular Formula: C15H21NO5
Formula Weight: 295.33
Physical Form: Solid
Melting Point: 58-60°
Beilstein Reference: 3064461
Optical Rotation: +20°(c=2 in ethanol)
MDL No.: MFCD00066063
Formula:
C15H21NO5 Melting Pt: 58…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066063 CAS Number: 23680-31-1 EINECS: 245-820-1 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...