Boc-N-Me-D-Phe-OH 98%
Supplier: Thermo Scientific Chemicals
H62638-03
H62638-06
H62638-MD
AAH62638-03EA
211.72
CAD
AAH62638-03
AAH62638-06
AAH62638-MD
Boc-N-Me-D-Phe-OH 98%
Boc-N-Me-D-Phe-OH
N-Boc-N-methyl-D-phenylalanine, 98%
Purity: 98%
CAS No.: 85466-66-6
Molecular Formula: C15H21NO4
Formula Weight: 279.33
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00151890
Formula:
C₁₅H₂₁NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00151890 CAS Number: 85466-66-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...