Boc-Ala-NH2 96%

Supplier: Thermo Scientific Chemicals

Synonyms: tert-butyl N-(1-carbamoylethyl)carbamate, Boc-Ala-NH₂, N-Boc-L-alaninamide

H62604-03 H62604-06 H62604-MD
AAH62604-03EA 278.08 CAD
AAH62604-03 AAH62604-06 AAH62604-MD
Boc-Ala-NH2 96%
Boc-Ala-NH2

N-Boc-L-alaninamide, 96%


Purity: 96% CAS No.: 85642-13-3 Molecular Formula: C8H16N2O3 Formula Weight: 188.22 Physical Form: Solid MDL No.: MFCD09263333

Formula: C₈H₁₆N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD09263333
CAS Number: 85642-13-3

Order Now

Specification Test Results

Assay 96%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR