Boc-Asp-OMe 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-Asp-OMe, N-Boc-L-aspartic acid 1-Methyl ester, N-Boc-L-aspartic acid 1-methyl ester, 1-Methyl N-Boc-L-aspartate, 1-Methyl N-(tert-Butoxycarbonyl)-L-aspartate, N-(tert-Butoxycarbonyl)-L-aspartic acid 1-Methyl ester

H62590-03 H62590-06
AAH62590-03EA 183.28 CAD
AAH62590-03 AAH62590-06 AAH62590-MD
Boc-Asp-OMe 98%
Boc-Asp-OMe

N-Boc-L-aspartic acid 1-methyl ester, 98%


Purity: 98% CAS No.: 98045-03-5 Molecular Formula: C10H17NO6 Formula Weight: 247.25 Physical Form: Solid MDL No.: MFCD03094777

Formula: C₁₀H₁₇NO₆
MW: 247.247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094777
CAS Number: 98045-03-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR