N,1-Di-Boc-L-histidine dicyclohexylammonium salt 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-His(Boc)-OH.DCHA, N,N'-Di-Boc-L-histidine dicyclohexylammonium salt, Boc-His(Boc)-OH·DCHA

H62570-06 H62570-14
AAH62570-06EA 84.53 CAD
AAH62570-06 AAH62570-14
N,1-Di-Boc-L-histidine dicyclohexylammonium salt 98%
N,1-Di-Boc-L-histidine dicyclohexylammonium salt

N,1-Di-Boc-L-histidine dicyclohexylammonium salt, 98%


Purity: 98% CAS No.: 31687-58-8 Molecular Formula: C16H25N3O6.C12H23N Formula Weight: 536.70 Physical Form: Solid Melting Point: 160-165° dec MDL No.: MFCD06804675

Formula: C₁₆H₂₅N₃O₆·C₁₂H₂₃N
MW: 536.714 g/mol
Melting Pt: 160…165 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD06804675
CAS Number: 31687-58-8
EINECS: 250-764-6

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR