Methyl (S)-(+)-2-pyrrolidone-5-carboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H62543-06
H62543-14
AAH62543-06EA
96.38
CAD
AAH62543-06
AAH62543-14
AAH62543-22
Methyl (S)-(+)-2-pyrrolidone-5-carboxylate 97%
Methyl (S)-(+)-2-pyrrolidone-5-carboxylate
Methyl (S)-(+)-2-oxopyrrolidine-5-carboxylate, 97%
Purity: 97%
CAS No.: 4931-66-2
Molecular Formula: C6H9NO3
Formula Weight: 143.14
Flash Point: 113°(235°F)
Physical Form: Liquid
Density: 1.226
Refractive Index: 1.4860
Appearance: Pale yellow
Boiling Point: 90°/0.3mm
Optical Rotation: +10°(c=1 in ethanol)
MDL No.: MFCD00080931
Formula:
C₆H₉NO₃ Boiling Pt: 90 °C/0.3 mm Density: 1.226 Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00080931 CAS Number: 4931-66-2 EINECS: 225-567-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...