trans-4-Cyanocyclohexylamine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (1r,4r)-4-aminocyclohexane-1-carbonitrile hydrochloride, trans-4-Aminocyclohexanecarbonitrile hydrochloride

H62522-03 H62522-MD
AAH62522-03EA 820.02 CAD
AAH62522-03 AAH62522-MD
trans-4-Cyanocyclohexylamine hydrochloride 97%
trans-4-Cyanocyclohexylamine hydrochloride

trans-4-Cyanocyclohexylamine hydrochloride, 97%


Purity: 97% CAS No.: 873537-33-8 Molecular Formula: C7H12N2.HCl Formula Weight: 160.64 UN No.: 3439 MDL No.: MFCD21643021

Formula: C₇H₁₃ClN₂
Storage Temperature: Ambient
MDL Number: MFCD21643021
CAS Number: 873537-33-8
UN: 3439
ADR: "6.1",III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR