L-(-)-Tryptophanol 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
L-Tryptophanol, (S)-(-)-2-Amino-3-(3-indolyl)propanol, (S)-2-Amino-3-(1H-indol-3-yl)propan-1-ol
H62506-03
H62506-06
AAH62506-03EA
167.48
CAD
AAH62506-03
AAH62506-06
L-(-)-Tryptophanol 97%
L-(-)-Tryptophanol
L-Tryptophanol, 97%
Purity: 97%
CAS No.: 2899-29-8
Molecular Formula: C11H14N2O
Formula Weight: 190.24
Physical Form: Viscous liquid
Appearance: Brown
Optical Rotation: -20° (c=1 in methanol)
MDL No.: MFCD00037970
Formula:
C₁₁H₁₄N₂O MW: 190.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00037970 CAS Number: 2899-29-8 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...