Boc-Glu(OMe)-OH 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-2-((tert-Butoxycarbonyl)amino)-5-methoxy-5-oxopentanoic acid, N-Boc-L-glutamic acid 5-methyl ester
H62496-03
H62496-06
H62496-MD
AAH62496-03EA
146.94
CAD
AAH62496-03
AAH62496-06
AAH62496-MD
Boc-Glu(OMe)-OH 95%
Boc-Glu(OMe)-OH
N-Boc-L-glutamic acid 5-methyl ester, 95%
Purity: 95%
CAS No.: 45214-91-3
Molecular Formula: C11H19NO6
Formula Weight: 261.27
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White
MDL No.: MFCD00190794
Formula:
C₁₁H₁₉NO₆ MW: 261.274 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00190794 CAS Number: 45214-91-3 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...