D-Alanine methyl ester hydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: D-Alanine Methyl Ester Hydrochloride, (R)-2-Aminopropionic acid Methyl Ester Hydrochloride, H-D-Ala-OMe.HCl, H-D-Ala-OMe·HCl, Methyl D-alaninate hydrochloride, Methyl (R)-2-Aminopropionate Hydrochloride, (R)-2-Aminopropionic Acid Methyl Ester Hydrochloride, (R)-Methyl 2-aminopropionate hydrochloride

H62472-06 H62472-14 H62472-22
AAH62472-06EA 58.93 CAD
AAH62472-06 AAH62472-14 AAH62472-22
D-Alanine methyl ester hydrochloride 98%
D-Alanine methyl ester hydrochloride

D-Alanine methyl ester hydrochloride, 98%


Purity: 98% CAS No.: 14316-06-4 Molecular Formula: C4H9NO2.HCl Formula Weight: 139.58 Physical Form: Crystalline powder Appearance: White to off-white Melting Point: 108-110° Beilstein Reference: 4235975 Optical Rotation: -8°(c=2 in methanol) MDL No.: MFCD00066141

Formula: C₄H₉NO₂
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00066141
CAS Number: 14316-06-4
EINECS: 238-258-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR