(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H62450-03
H62450-06
AAH62450-03EA
78.37
CAD
AAH62450-03
AAH62450-06
AAH62450-MD
(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid 97%
(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid
(S)-4-Amino-2-(Boc-amino)butyric acid, 97%
Purity: 97%
CAS No.: 25691-37-6
Molecular Formula: C9H18N2O4
Formula Weight: 218.25
Physical Form: Powder
Appearance: White to off-white
Beilstein Reference: 4745487
Optical Rotation: +7°(c=1 in methanol)
MDL No.: MFCD00236841
Formula:
C₉H₁₈N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00236841 CAS Number: 25691-37-6 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...