H-ORN(Z)-OH 98%

Supplier: Thermo Scientific Chemicals

H62435-03 H62435-06 H62435-14
AAH62435-03EA 59.25 CAD
AAH62435-03 AAH62435-06 AAH62435-14
H-ORN(Z)-OH 98%
H-ORN(Z)-OH

N^d-Benzyloxycarbonyl-L-ornithine, 98%


Purity: 98% CAS No.: 3304-51-6 Molecular Formula: C13H18N2O4 Formula Weight: 266.29 Physical Form: Solid MDL No.: MFCD00038200

Formula: C₁₃H₁₈N₂O₄
Melting Pt: 248…252 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037220
CAS Number: 3304-51-6
EINECS: 221-980-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR