Boc-Asp-OtBu 95%
Supplier: Thermo Scientific Chemicals
H62425-03
H62425-06
AAH62425-03EA
89.27
CAD
AAH62425-03
AAH62425-06
Boc-Asp-OtBu 95%
Boc-Asp-OtBu
N-Boc-L-aspartic acid 1-tert-butyl ester, 95%
Purity: 95%
CAS No.: 34582-32-6
Molecular Formula: C13H23NO6
Formula Weight: 289.32
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Odor: Odorless
Beilstein Reference: 4191701
Optical Rotation: -24°(c=1 in methanol)
MDL No.: MFCD00038272
Formula:
C₁₃H₂₃NO₆ Storage Temperature: Ambient |
MDL Number:
MFCD00038272 CAS Number: 34582-32-6 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...