Fmoc-D-Dap(Boc)-OH 95%
Supplier: Thermo Scientific Chemicals
H62381-03
H62381-06
AAH62381-03EA
222.78
CAD
AAH62381-03
AAH62381-06
AAH62381-MD
Fmoc-D-Dap(Boc)-OH 95%
Fmoc-D-Dap(Boc)-OH
(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 95%
Purity: 95%
CAS No.: 198544-42-2
Molecular Formula: C23H26N2O6
Formula Weight: 426.46
Physical Form: Solid
MDL No.: MFCD00798648
Formula:
C₂₃H₂₆N₂O₆ Storage Temperature: Ambient |
MDL Number:
MFCD00798648 CAS Number: 198544-42-2 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...