Boc-D-Glu-OtBu 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Boc-D-glutamic acid 1-tert-butyl ester
H62320-03
H62320-06
AAH62320-03EA
290.72
CAD
AAH62320-03
AAH62320-06
AAH62320-MD
Boc-D-Glu-OtBu 95%
Boc-D-Glu-OtBu
N-Boc-D-glutamic acid 1-tert-butyl ester, 95%
Purity: 95%
CAS No.: 73872-71-6
Molecular Formula: C14H25NO6
Formula Weight: 303.35
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale brown
MDL No.: MFCD00076926
Formula:
C₁₄H₂₅NO₆ Storage Temperature: Ambient |
MDL Number:
MFCD00076926 CAS Number: 73872-71-6 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...